p-Xylylendiamin

Lieferant: ABCR

Synonyme: 1,4-Bis(aminomethyl)benzene, 1,4-Phenylenedimethanamine

AB143306.0025
ABCRAB143306.0025EA 0 EUR
ABCRAB143306.0025
p-Xylylendiamin
p-Xylylendiamin
Formel: C₈H₁₂N₂
Molecular Weight: 136,20 g/mol
MDL: MFCD00009821
CAS-Nummer: 539-48-0
EINECS: 208-719-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR