1-(2-Pyridyl)piperazin
Lieferant: ABCR
Synonyme:
2-(1-Piperazinyl)pyridin
AB143546.0025
ABCRAB143546.0025EA
0
EUR
ABCRAB143546.0025
1-(2-Pyridyl)piperazin
1-(2-Pyridyl)piperazin
Formel:
C₉H₁₃N₃ Molecular Weight: 163,22 g/mol Siedepunkt: 293 °C (1013 hPa) Schmelzpunkt: 85…87 °C Dichte: 1,07 g/cm³ (20 °C) |
MDL:
MFCD00006216 CAS-Nummer: 34803-66-2 EINECS: 252-220-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...