5-Fluor-2-methylbenzaldehyd

Lieferant: ABCR

AB144145.0005
ABCRAB144145.0005EA 0 EUR
ABCRAB144145.0005
5-Fluor-2-methylbenzaldehyd
5-Fluor-2-methylbenzaldehyd
Formel: C₈H₇FO
Molecular Weight: 138,14 g/mol
MDL: MFCD02261763
CAS-Nummer: 22062-53-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR