Girard-Reagenz D (N,N-Dimethylglycinhydrazid Monohydrochlorid)

Lieferant: ABCR

AB144158.0010
ABCRAB144158.0010EA 0 EUR
ABCRAB144158.0010
Girard-Reagenz D (N,N-Dimethylglycinhydrazid Monohydrochlorid)
Girard-Reagenz D (N,N-Dimethylglycinhydrazid Monohydrochlorid)
Formel: C₄H₁₂ClN₃O
CAS-Nummer: 539-64-0
EINECS: 208-721-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR