2,4-Dimethylpyrrol

Lieferant: ABCR

Synonyme: 2,4-Dimethyl-1H-pyrrole

AB144191.0005
ABCRAB144191.0005EA 0 EUR
ABCRAB144191.0005
2,4-Dimethylpyrrol
2,4-Dimethylpyrrole
Formel: C₆H₉N
Molecular Weight: 95,1424 g/mol
MDL: MFCD00192088
CAS-Nummer: 625-82-1
EINECS: 210-912-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR