8-(Brommethyl)chinolin

Lieferant: abcr

AB145350.0250
ABCRAB145350.0250EA 0 EUR
ABCRAB145350.0250
8-(Brommethyl)chinolin
8-(Bromomethyl)quinoline
Formel: C₁₀H₈BrN
MDL: MFCD00047621
CAS-Nummer: 7496-46-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR