3-(Chlormethyl)-5-methylisoxazol 95%

Lieferant: ABCR

AB145365.0001
ABCRAB145365.0001EA 0 EUR
ABCRAB145365.0001
3-(Chlormethyl)-5-methylisoxazol 95%
3-(Chloromethyl)-5-methylisoxazole
Formel: C₅H₆ClNO
Molecular Weight: 131,560 g/mol
MDL: MFCD00221166
CAS-Nummer: 35166-37-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR