2-Aminochinolin
Lieferant: ABCR
Synonyme:
Quinolin-2-ylamine
AB147659.0250
ABCRAB147659.0250EA
0
EUR
ABCRAB147659.0250
2-Aminochinolin
2-Aminochinolin
Formel:
C₉H₈N₂ Molecular Weight: 144,18 g/mol Siedepunkt: 305 °C (1013 hPa) Schmelzpunkt: 130 °C |
MDL:
MFCD00038079 CAS-Nummer: 580-22-3 EINECS: 209-458-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...