4-Cyan-2,3,5,6-tetrafluorbenzotrifluorid

Lieferant: ABCR

Synonyme: 2,3,5,6-Tetrafluoro-4-(trifluoromethyl)benzonitrile

AB148414.0010
ABCRAB148414.0010EA 0 EUR
ABCRAB148414.0010
4-Cyan-2,3,5,6-tetrafluorbenzotrifluorid
4-Cyan-2,3,5,6-tetrafluorbenzotrifluorid
Formel: C₈F₇N
MDL: MFCD04972776
CAS-Nummer: 15895-68-8
UN: 3276
ADR: 6.1,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR