4-Fluor-3-methoxybenzylbromid

Lieferant: ABCR

Synonyme: 4-(Bromomethyl)-1-fluoro-2-methoxybenzene

AB148462.0010
ABCRAB148462.0010EA 0 EUR
ABCRAB148462.0010
4-Fluor-3-methoxybenzylbromid
4-Fluor-3-methoxybenzylbromid
Formel: C₈H₈BrFO
Molecular Weight: 219,0509 g/mol
MDL: MFCD04972878
CAS-Nummer: 141080-73-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR