1,3,5-Trimethyl-1H-pyrazol-4-sulfonylchlorid

Lieferant: ABCR

Synonyme: 1,3,5-Trimethyl-1H-pyrazole-4-sulphonyl chloride

AB148721.0001
ABCRAB148721.0001EA 0 EUR
ABCRAB148721.0001
1,3,5-Trimethyl-1H-pyrazol-4-sulfonylchlorid
1,3,5-Trimethyl-1H-pyrazol-4-sulfonylchlorid
Formel: C₆H₉ClN₂O₂S
Molecular Weight: 208,6700 g/mol
MDL: MFCD02180944
CAS-Nummer: 59340-27-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR