(6-Morpholino-3-pyridinyl)methanol

Lieferant: ABCR

CC17509CB CC17509DA AB149355.0001
MAYBCC17509CBEA 0 EUR
MAYBCC17509CB MAYBCC17509DA ABCRAB149355.0001
(6-Morpholino-3-pyridinyl)methanol
(6-Morpholino-3-pyridinyl)methanol
Formel: C₁₀H₁₄N₂O₂
MDL: MFCD03086115
CAS-Nummer: 388088-73-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR