4-(Methylthio)benzylamin

Lieferant: ABCR

Synonyme: (4-(Methylthio)phenyl)methanamine

AB149996.0005
ABCRAB149996.0005EA 423 EUR
ABCRAB149996.0005
4-(Methylthio)benzylamin
4-(Methylthio)benzylamin
Formel: C₈H₁₁NS
Molecular Weight: 153,2500 g/mol
MDL: MFCD01310831
CAS-Nummer: 83171-39-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR