4-Aminopyrimidin

Lieferant: ABCR
Achtung

AB151597.0005
ABCRAB151597.0005EA 0 EUR
ABCRAB151597.0005
4-Aminopyrimidin
4-Aminopyrimidin
Formel: C₄H₅N₃
Molecular Weight: 95,1 g/mol
Schmelzpunkt: 154…156 °C
MDL: MFCD00006112
CAS-Nummer: 591-54-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR