6-Brom-2,2'-bipyridyl

Lieferant: ABCR

Synonyme: 6-Bromo-2,2'-bipyridine

AB152211.0010
ABCRAB152211.0010EA 0 EUR
ABCRAB152211.0010
6-Brom-2,2'-bipyridyl
6-Brom-2,2'-bipyridyl
Formel: C₁₀H₇BrN₂
Molecular Weight: 235,0800 g/mol
MDL: MFCD01318953
CAS-Nummer: 10495-73-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR