Ethyltrichlorsilan
Lieferant: abcr
Synonyme:
Ethylsilyltrichlorid, Trichlor(ethyl)silan
AB153238.0025
ABCRAB153238.0025EA
0
EUR
ABCRAB153238.0025
Ethyltrichlorsilan
Ethyltrichlorsilan
Formel:
C₂H₅Cl₃Si Molecular Weight: 163.51 g/mol Siedepunkt: 102 Schmelzpunkt: –105.6 °C Dichte: 1.24 g/cm³ (20 °C) |
MDL:
MFCD00000485 CAS-Nummer: 115-21-9 EINECS: 204-072-6 UN: 1196 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...