Dimethoxy(dimethyl)silan

Lieferant: ABCR

Synonyme: Dimethyldimethoxysilan

AB153326.0500
ABCRAB153326.0500EA 0 EUR
ABCRAB153326.0500
Dimethoxy(dimethyl)silan
Dimethoxy(dimethyl)silan
Formel: C₄H₁₂O₂Si
Molecular Weight: 120,22 g/mol
Siedepunkt: 82 °C (1013 hPa)
Schmelzpunkt: –80 °C
Dichte: 0,8638 g/cm³ (20 °C)
MDL: MFCD00025691
CAS-Nummer: 1112-39-6
EINECS: 214-189-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR