Methylimidazo[1,2-a]pyridin-6-carboxylat

Lieferant: ABCR

AB153896.0500
ABCRAB153896.0500EA 0 EUR
ABCRAB153896.0500
Methylimidazo[1,2-a]pyridin-6-carboxylat
Methylimidazo[1,2-a]pyridin-6-carboxylat
Formel: C₉H₈N₂O₂
Molecular Weight: 176,1720 g/mol
MDL: MFCD06200984
CAS-Nummer: 136117-69-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR