DL-Phenylalaninol

Lieferant: ABCR

Synonyme: 2-Amino-3-phenyl-1-propanol, DL-2-Amino-3-phenyl-1-propanol, 2-Amino-3-phenylpropan-1-ol

AB153943.0005
ABCRAB153943.0005EA 0 EUR
ABCRAB153943.0005
DL-Phenylalaninol
DL-Phenylalaninol
Formel: C₉H₁₃NO
Molecular Weight: 151,21 g/mol
MDL: MFCD00066689
CAS-Nummer: 16088-07-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR