6-Methoxyindol

Lieferant: ABCR
Achtung

Synonyme: 6-Methoxy-1H-indol, Indol-6-ylmethylether

AB168523.0005
ABCRAB168523.0005EA 0 EUR
ABCRAB168523.0005
6-Methoxyindol
6-Methoxyindol
Formel: C₉H₉NO
Molecular Weight: 147,18 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00022780
CAS-Nummer: 3189-13-7
EINECS: 221-689-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR