(S)-(+)-β-Aminophenethylalkohol
Lieferant: ABCR
Synonyme:
(S)-(+)-2-Phenylglycinol, L-Phenylglycinol, (S)-2-Amino-2-phenylethanol
AB168529.0025
ABCRAB168529.0025EA
0
EUR
ABCRAB168529.0025
(S)-(+)-β-Aminophenethylalkohol
(S)-(+)-β-Aminophenethylalkohol
Formel:
C₈H₁₁NO Molecular Weight: 137,18 g/mol |
MDL:
MFCD00064404 CAS-Nummer: 20989-17-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...