3-Fluoranthranilsäure

Lieferant: ABCR

Synonyme: 2-Amino-3-fluorobenzoic acid

AB168567-0005
ABCRAB168567-0005EA 0 EUR
ABCRAB168567-0005
3-Fluoranthranilsäure
3-Fluoranthranilsäure
Formel: C₇H₆FNO₂
Molecular Weight: 155,13 g/mol
MDL: MFCD01569395
CAS-Nummer: 825-22-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR