5-Fluorisatosäureanhydrid

Lieferant: ABCR

Synonyme: 6-Fluoro-2H-benzo[d][1,3]oxazine-2,4(1H)-dione

AB170442.0005
ABCRAB170442.0005EA 0 EUR
ABCRAB170442.0005
5-Fluorisatosäureanhydrid
5-Fluorisatosäureanhydrid
Formel: C₈H₄FNO₃
Molecular Weight: 181,12 g/mol
MDL: MFCD03300719
CAS-Nummer: 321-69-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR