2-Pyridinboronsäure

Lieferant: ABCR

Synonyme: Pyridin-2-boronsäure, 2-Pyridinylboronsäure

AB170673.0100
ABCRAB170673.0100EA 0 EUR
ABCRAB170673.0100
2-Pyridinboronsäure
2-Pyridineboronic acid
Formel: C₅H₆BNO₂
Molecular Weight: 122,92 g/mol
MDL: MFCD00151844
CAS-Nummer: 197958-29-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR