3-Amino-4-methylpyridin 98%

Lieferant: ABCR
Gefahr

AB172495.0005
ABCRAB172495.0005EA 0 EUR
ABCRAB172495.0005
3-Amino-4-methylpyridin 98%
3-Amino-4-methylpyridin

Purity: 98% CAS No.: 3430-27-1 Molecular Formula: C6H8N2 Formula Weight: 108.14 Odour: Unpleasant Melting Point: 102-106° Boiling Point: 254° Beilstein Reference: 107792 UN No.: 2811 MDL No.: MFCD00128871

Formel: C₆H₈N₂
Molecular Weight: 108,14 g/mol
Siedepunkt: 254 °C
Schmelzpunkt: 102…106 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00128871
CAS-Nummer: 3430-27-1
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR