4-Methoxy-2,2,6,6-tetramethylpiperidin-1-oxyl (4-Methoxy-TEMPO) ≥98% freies Radikal

Lieferant: ABCR
Achtung

Synonyme: 4-Methoxy-2,2,6,6-tetramethylpiperidine 1-Oxyl, 4-Methoxy-2,2,6,6-tetramethyl-1-piperidinyloxy

AB173085.0001
ABCRAB173085.0001EA 0 EUR
ABCRAB173085.0001
4-Methoxy-2,2,6,6-tetramethylpiperidin-1-oxyl (4-Methoxy-TEMPO) ≥98% freies Radikal
4-Methoxy-2,2,6,6-tetramethylpiperidin-1-oxyl (4-Methoxy-TEMPO)

Purity: 98+% CAS No.: 95407-69-5 Molecular Formula: C10H20NO2 Formula Weight: 186.28 Melting Point: 39-41° Beilstein Reference: 4740399

Formel: C₁₀H₂₀NO₂
Molecular Weight: 186,27 g/mol
Schmelzpunkt: 39…41 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00270334
CAS-Nummer: 95407-69-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR