(S)-(-)-4-Brom-α-methylbenzylamin 99%, ee 98%, ChiPros®

Lieferant: ABCR
Gefahr

Synonyme: (S)-1-(4-Bromophenyl)ethanamine, (S)-(-)-1-(4-Bromophenyl)ethylamine

AB173493.0005
ABCRAB173493.0005EA 0 EUR
ABCRAB173493.0005
(S)-(-)-4-Brom-α-methylbenzylamin 99%, ee 98%, ChiPros®
(S)-(-)-4-Brom-α-methylbenzylamin

Purity: 99% CAS No.: 27298-97-1 Molecular Formula: C8H10BrN Formula Weight: 200.08 Flash Point: 110°(230°F) Refractive Index: 1.5670 Melting Point: -25° (neat) Boiling Point: 140-145°/30mm Beilstein Reference: 5729988 UN No.: 2735 MDL No.: MFCD00066026

Formel: C₈H₁₀BrN
Molecular Weight: 200,08 g/mol
Siedepunkt: 140…145 °C (30 mmHg)
Schmelzpunkt: –25 °C
Dichte: 1,371
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00066026
CAS-Nummer: 27298-97-1
UN: 2735
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR