3-Formylphenylboronsäure

Lieferant: ABCR

Synonyme: 3-Boronobenzaldehyd, Dihydroxy(3-formylphenyl)boran, (3-Formylphenyl)metaborsäure, 3-Formylbenzolboronsäure

AB175246.0005
ABCRAB175246.0005EA 0 EUR
ABCRAB175246.0005
3-Formylphenylboronsäure
3-Formylphenylboronsäure
Formel: C₇H₇BO₃
Molecular Weight: 149,94 g/mol
MDL: MFCD00161356
CAS-Nummer: 87199-16-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR