4-Propylanilin ≥98%

Lieferant: ABCR
Gefahr

AB177792.0025
ABCRAB177792.0025EA 0 EUR
ABCRAB177792.0025
4-Propylanilin ≥98%
4-Propylanilin

Purity: 98% CAS No.: 2696-84-6 Molecular Formula: C9H13N Formula Weight: 135.21 Flash Point: 103°(217°F) Refractive Index: 1.5430 Boiling Point: 224-226° Beilstein Reference: 2205524 UN No.: 2810 MDL No.: MFCD00007924

Formel: C₉H₁₃N
Molecular Weight: 135,21 g/mol
Siedepunkt: 224…226 °C
Dichte: 0,919
Flammpunkt: 103 °C (217 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007924
CAS-Nummer: 2696-84-6
EINECS: 220-271-0
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR