N(α)-Boc-D-tryptophan ≥97%
Lieferant: Thermo Fisher Scientific
Synonyme:
Boc-D-Trp-OH
L09214.03
L09214.06
AB178718.0005
L09214.03EA
20.8
EUR
L09214.03
L09214.06
ABCRAB178718.0005
N(α)-Boc-D-tryptophan ≥97%
N(α)-Boc-D-tryptophan
Purity: 97%
CAS No.: 5241-64-5
Molecular Formula: C16H20N2O4
Formula Weight: 304.35
Storage Temperature: 2°C to 8°C
Melting Point: ca 138° dec.
Beilstein Reference: 4237334
Optical Rotation: +21° (c=1 in acetic acid)
MDL No.: MFCD00037944
Formel:
C₁₆H₂₀N₂O₄ Molecular Weight: 304,3 g/mol Schmelzpunkt: ∼ 138 °C (Zersetzung) Lagertemperatur: Kühlschrank |
MDL:
MFCD00037944 CAS-Nummer: 5241-64-5 EINECS: 226-042-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...