3,3-Dimethyl-1,5-dioxaspiro[5.5]undecan-9-on 95%

Lieferant: ABCR

Synonyme: 1,4-Cyclohexanedione mono(2,2-dimethyltrimethylene ketal)

AB178783.0025
ABCRAB178783.0025EA 0 EUR
ABCRAB178783.0025
3,3-Dimethyl-1,5-dioxaspiro[5.5]undecan-9-on 95%
3,3-Dimethyl-1,5-dioxaspiro[5.5]undecan-9-on

Purity: 95% CAS No.: 69225-59-8 Molecular Formula: C11H18O3 Formula Weight: 198.26 Melting Point: 46-50° Beilstein Reference: 3541930

Formel: C₁₁H₁₈O₃
Molecular Weight: 198,26 g/mol
Schmelzpunkt: 46…50 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006652
CAS-Nummer: 69225-59-8
EINECS: 273-918-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR