2-Methyl-3-nitrophenylisocyanat ≥98%

Lieferant: ABCR

Synonyme: 2-Isocyanato-6-nitrotoluene, 1-Isocyanato-2-methyl-3-nitrobenzene

AB179055.0005
ABCRAB179055.0005EA 0 EUR
ABCRAB179055.0005
2-Methyl-3-nitrophenylisocyanat ≥98%
2-Methyl-3-nitrophenylisocyanat

Purity: 98% CAS No.: 23695-15-0 Molecular Formula: C8H6N2O3 Formula Weight: 178.15 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Odour: Irritating Melting Point: 34-37° Boiling Point: 118°/2.8mm Beilstein Reference: 9120119 UN No.: 2811 MDL No.: MFCD00037046

Formel: C₈H₆N₂O₃
Molecular Weight: 178,15 g/mol
Siedepunkt: 118 °C (2,8 mmHg)
Schmelzpunkt: 34…37 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Kühlschrank
MDL: MFCD00037046
CAS-Nummer: 23695-15-0
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR