Methyl-2-isocyanatobenzoat ≥97%
Lieferant: ABCR
Synonyme:
2-(Methoxycarbonyl)phenyl isocyanate
AB179489.0005
ABCRAB179489.0005EA
0
EUR
ABCRAB179489.0005
Methyl-2-isocyanatobenzoat ≥97%
Methyl-2-isocyanatobenzoat
Purity: 97%
CAS No.: 1793-07-3
Molecular Formula: C9H7NO3
Formula Weight: 177.16
Storage Temperature: 2°C to 8°C
Odour: Irritating
Boiling Point: 145-146°/16mm
Beilstein Reference: 1241370
UN No.: 2811
MDL No.: MFCD00013848
Formel:
C₉H₇NO₃ Schmelzpunkt: 45…49 °C |
MDL:
MFCD00013848 CAS-Nummer: 1793-07-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...