ω-Acetoxyacetophenon ≥98%

Lieferant: ABCR
Achtung

Synonyme: Phenacyl acetate, Acetic acid phenacyl ester, 2-Acetoxy-1-phenylethan-1-one, 1-(2-Acetoxyphenyl)ethanone, 2-Acetoxyacetophenone

AB180041.0025
ABCRAB180041.0025EA 0 EUR
ABCRAB180041.0025
ω-Acetoxyacetophenon ≥98%
ω-Acetoxyacetophenon

Purity: 98% CAS No.: 2243-35-8 Molecular Formula: C10H10O3 Formula Weight: 178.19 Flash Point: 110°(230°F) Melting Point: 48-50° Boiling Point: 110-114°/2mm Beilstein Reference: 1240545 MDL No.: MFCD00026205

Formel: C₆H₅C(O)CH₂OC(O)CH₃
Molecular Weight: 178,19 g/mol
Schmelzpunkt: 48…50 °C
Flammpunkt: >110 °C (230 °F)
MDL: MFCD00026205
CAS-Nummer: 2243-35-8
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=56de6ff41a047fce58a9dfa08532c69410ff35bd&vid=56de6ff41a047fce58a9dfa08532c69410ff35bd&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR