4-tert-Butoxybenzonitril 96%
Lieferant: abcr
|
Achtung
|
AB180471.0025
ABCRAB180471.0025EA
0
EUR
ABCRAB180471.0025
4-tert-Butoxybenzonitril 96%
4-tert-Butoxybenzonitril
Purity: 96%
CAS No.: 185259-36-3
Molecular Formula: C11H13NO
Formula Weight: 175.23
Refractive Index: 1.5210
Boiling Point: 95°/0.8mm
MDL No.: MFCD03425273
Formel:
C₁₁H₁₃NO Siedepunkt: 95 °C (0,8 mmHg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD03425273 CAS-Nummer: 185259-36-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...