(S)-(+)-4-Methyl-2-pentanol ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(S)-(+)-Methylisobutylcarbinol
L18882.03
L18882.MD
AB180566.0001
L18882.03EA
144
EUR
L18882.03
L18882.MD
ABCRAB180566.0001
(S)-(+)-4-Methyl-2-pentanol ≥99%
(S)-(+)-4-Methyl-2-pentanol
Purity: 99%
CAS No.: 14898-80-7
Molecular Formula: C6H14O
Formula Weight: 102.18
Flash Point: 41°(105°F)
Refractive Index: 1.4110
Odour: Slightly sweetish
Boiling Point: 130-131°
Beilstein Reference: 1718991
Optical Rotation: +21° (neat)
UN No.: 2053
MDL No.: MFCD03093078
Formel:
C₆H₁₄O Molecular Weight: 102.18 g/mol Siedepunkt: 130…131 °C Dichte: 0.802 Flammpunkt: 41 °C (105 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD03093078 CAS-Nummer: 14898-80-7 UN: 2053 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...