4-(Brommethyl)-3-fluorbenzonitril

Lieferant: ABCR

AB200602-5G
ABCRAB200602-5GEA 0 EUR
ABCRAB200602-5G
4-(Brommethyl)-3-fluorbenzonitril
4-(Brommethyl)-3-fluorbenzonitril
Formel: C₈H₅BrFN
Molecular Weight: 214,04 g/mol
MDL: MFCD07368341
CAS-Nummer: 105942-09-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR