trans-Bis(triphenylphosphin)palladium(II)chlorid ≥99,95% (Metall-Basis) Pd ≥ 14.7%, Premion®

Lieferant: abcr

Synonyme: trans-Dichlorobis(triphenylphosphine)palladium(II)

AB209725.0005
ABCRAB209725.0005EA 0 EUR
ABCRAB209725.0005
trans-Bis(triphenylphosphin)palladium(II)chlorid ≥99,95% (Metall-Basis) Pd ≥ 14.7%, Premion®
trans-Bis(triphenylphosphin)palladium(II)chlorid
Formel: C₃₆H₃₀Cl₂P₂Pd
Molecular Weight: 701,91 g/mol
Schmelzpunkt: ∼ 310 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009593
CAS-Nummer: 13965-03-2
EINECS: 237-744-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR