Antimon(III)sulfid
Lieferant: ABCR
|
Achtung
|
Synonyme:
di-Antimontrisulfid, Antimontrisulfid
AV12663-1000
ABCRAV12663-1000EA
0
EUR
ABCRAV12663-1000
Antimon(III)sulfid
Antimon(III)sulfid
Formel:
S₃Sb₂ Molecular Weight: 339,72 g/mol Siedepunkt: 1150 °C (1013 hPa) Schmelzpunkt: 550 °C Dichte: 4,12 g/cm³ (20 °C) |
MDL:
MFCD00011217 CAS-Nummer: 1345-04-6 EINECS: 215-713-4 UN: 1549 ADR: 6.1,III Merck Index: 12,00754 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...