Thallium(III)nitrat

Lieferant: ABCR

AV17399.0025
ABCRAV17399.0025EA 0 EUR
ABCRAV17399.0025
Thallium(III)nitrat
Thallium(III)nitrat
Formel: N₃O₉Tl
Molecular Weight: 390,4 g/mol
MDL: MFCD00011272
CAS-Nummer: 13746-98-0
EINECS: 237-325-4
UN: 2727
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR