4-(Aminomethyl)tetrahydropyran

Lieferant: ABCR

Synonyme: 4-(Aminomethyl)tetrahydro-2H-pyran, 4-Aminomethyltetrahydropyran, Tetrahydro-2H-pyran-4-ylmethanamine, (Tetrahydro-2H-pyran-4-yl)methanamine

CAM-1003.0005
ABCRCAM-1003.0005EA 0 EUR
ABCRCAM-1003.0005
4-(Aminomethyl)tetrahydropyran
4-(Aminomethyl)tetrahydropyran
Formel: C₆H₁₃NO
Molecular Weight: 115,1735 g/mol
MDL: MFCD02179435
CAS-Nummer: 130290-79-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR