Tetrahydropyran-4-carbonsäure

Lieferant: ABCR

Synonyme: Tetrahydropyran-4-yl-carboxylic acid

CAM-1006
ABCRCAM-1006EA 0 EUR
ABCRCAM-1006
Tetrahydropyran-4-carbonsäure
Tetrahydropyran-4-carbonsäure
Formel: C₆H₁₀O₃
Molecular Weight: 130,14 g/mol
MDL: MFCD00031016
CAS-Nummer: 5337-03-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR