2-Bromfuran

Lieferant: Thermo Scientific

CC02010.250 CC02010DA CC02010.0001
MAYBCC02010.250EA 0 EUR
MAYBCC02010.250 MAYBCC02010DA ABCRCC02010.0001
2-Bromfuran
2-Bromofuran
Formel: C₄H₃BrO
MDL: MFCD00216609
CAS-Nummer: 584-12-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR