2,1,3-Benzothiadiazol-5-carbonsäure

Lieferant: abcr

CC09201CB CC09201DA CC09201.0001
MAYBCC09201CBEA 0 EUR
MAYBCC09201CB MAYBCC09201DA ABCRCC09201.0001
2,1,3-Benzothiadiazol-5-carbonsäure
2,1,3-Benzothiadiazole-5-carboxylic acid
Formel: C₇H₄N₂O₂S
MDL: MFCD01647555
CAS-Nummer: 16405-98-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR