7-Iod-3,4-dihydro-2H-1,5-benzodioxepin

Lieferant: Thermo Scientific

CC13211.250 CC13211DA CC13211.0001
MAYBCC13211.250EA 0 EUR
MAYBCC13211.250 MAYBCC13211DA ABCRCC13211.0001
7-Iod-3,4-dihydro-2H-1,5-benzodioxepin
7-Iod-3,4-dihydro-2H-1,5-benzodioxepin
Formel: C₉H₉IO₂
MDL: MFCD02677706
CAS-Nummer: 306934-90-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR