4-Amino-3-(trifluormethoxy)benzoesäure

Lieferant: abcr

F01100BD.0001
ABCRF01100BD.0001EA 0 EUR
ABCRF01100BD.0001
4-Amino-3-(trifluormethoxy)benzoesäure
4-Amino-3-(trifluoromethoxy)benzoic acid
Formel: C₈H₆F₃NO₃
MDL: MFCD00205143
CAS-Nummer: 175278-22-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR