Bis(2,2,2-trifluorethyl)amin

Lieferant: ABCR

F01253.0001
ABCRF01253.0001EA 0 EUR
ABCRF01253.0001
Bis(2,2,2-trifluorethyl)amin
Bis(2,2,2-trifluorethyl)amin
Formel: C₄H₅F₆N
Molecular Weight: 181,08 g/mol
MDL: MFCD00042094
CAS-Nummer: 407-01-2
EINECS: 206-978-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR