4-Brom-2,2-difluor-1,3-benzodioxol

Lieferant: ABCR
Achtung

Synonyme: 4-Bromo-2,2-difluorobenzo[d][1,3]dioxole

F01385G.0001
ABCRF01385G.0001EA 0 EUR
ABCRF01385G.0001
4-Brom-2,2-difluor-1,3-benzodioxol
4-Brom-2,2-difluor-1,3-benzodioxol
Formel: C₇H₃BrF₂O₂
Molecular Weight: 237 g/mol
MDL: MFCD01631385
CAS-Nummer: 144584-66-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR