3-Brom-4-(trifluormethoxy)anilin

Lieferant: ABCR

F01593KA.0010
ABCRF01593KA.0010EA 0 EUR
ABCRF01593KA.0010
3-Brom-4-(trifluormethoxy)anilin
3-Brom-4-(trifluormethoxy)anilin
Formel: C₇H₅BrF₃NO
Molecular Weight: 256,02 g/mol
MDL: MFCD00190127
CAS-Nummer: 191602-54-7
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR