2-Brom-5-(trifluormethyl)pyridin

Lieferant: ABCR

F01598G
ABCRF01598GEA 0 EUR
ABCRF01598G
2-Brom-5-(trifluormethyl)pyridin
2-Bromo-5-(trifluoromethyl)pyridine
Formel: C₆H₃BrF₃N
Molecular Weight: 226 g/mol
MDL: MFCD00153086
CAS-Nummer: 50488-42-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR